Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KIFC3 SIRNA Assays ›
          Gene Symbol:
          KIFC3
          Gene Name:
          kinesin family member C3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 57758217 - 57863053 on Build GRCh38
          UniGene ID:
          Hs.23131
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s7845

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KIFC3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130099.1 NP_001123571.1 16 2414
          NM_001130100.1 NP_001123572.1 17 2470
          NM_001318710.1 NP_001305639.1 17 2351
          NM_001318711.1 NP_001305640.1 17 2512
          NM_001318712.1 NP_001305641.1 15 2087
          NM_001318713.1 NP_001305642.1 15 2480
          NM_001318714.1 NP_001305643.1 18 2859
          NM_001318715.1 NP_001305644.1 17 2448
          NM_005550.3 NP_005541.3 17 2470
          NR_134678.1 17 2752
          XM_005255937.1 XP_005255994.1 19 2979
          XM_005255944.1 XP_005256001.1 17 2333
          XM_006721188.1 XP_006721251.1 20 3057
          XM_011523075.1 XP_011521377.1 17 2637
          XM_011523076.1 XP_011521378.1 17 2637
          XM_011523077.1 XP_011521379.1 17 2436
          XM_011523078.1 XP_011521380.1 17 2239
          XM_011523079.1 XP_011521381.1 17 2574
          XM_017023219.1 XP_016878708.1 20 3026
          XM_017023220.1 XP_016878709.1 20 3203
          XM_017023221.1 XP_016878710.1 19 2979
          XM_017023222.1 XP_016878711.1 21 3221
          XM_017023223.1 XP_016878712.1 17 2354
          XM_017023224.1 XP_016878713.1 19 2888
          XM_017023225.1 XP_016878714.1 15 3458
          XM_017023226.1 XP_016878715.1 18 2731
          XM_017023227.1 XP_016878716.1 18 2731
          GenBank mRNA AB209290.1 - 17 2405
          AF004426.1 - 13 1856
          AK091891.1 - 2236
          AK291737.1 - 16 2309
          AK295926.1 - 12 1432
          AK296995.1 - 17 2350
          AK299431.1 - 17 2412
          AK302743.1 - 15 2087
          AK303021.1 - 15 2408
          AK304132.1 - 17 2458
          AK307846.1 - 17 2428
          BC001211.1 - 17 2439 3503847
          BC008014.1 - 11 1435
          BC019057.2 - 4 439
          BC034234.1 - 18 2799 5296397
          BC041132.1 - 17 2300 5760988
          BC047051.1 - 17 2696 5297265
          BX647698.1 - 2 75

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          microtubule binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule-based movement
          Golgi organization
          visual perception
          zonula adherens maintenance
          epithelial cell-cell adhesion
          microtubule motor activity
          protein binding
          ATP binding
          microtubule binding
          ATP-dependent microtubule motor activity, minus-end-directed
          ATPase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0