Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015810.2 | NP_056625.2 | 1 | 332 | ||
GenBank mRNA | AF177202.1 | - | 1 | 237 | ||
AK041591.1 | - | 1 | 304 | |||
AK042777.1 | - | 1 | 268 | |||
AK164730.1 | - | 1 | 315 | |||
AK169977.1 | - | 1 | 334 | |||
BC119131.1 | - | 1 | 261 | 8733880 | ||
BC144810.1 | - | 1 | 261 |