Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KIR2DS1 SIRNA Assays ›
          Gene Symbol:
          KIR2DS1 KIR2DS2 KIR2DS4 KIR3DS1
          Gene Name:
          killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 1
          killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 2
          killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4
          killer cell immunoglobulin like receptor, three Ig domains and short cytoplasmic tail 1
          Gene Aliases:
          CD158H, CD158a, KIR2DP1DL1, KIR2DS4, p50.1
          183ActI, CD158J, CD158b, KIR-2DS2, NKAT-5, NKAT5, cl-49
          CD158I, KIR-2DS4, KIR1D, KIR2DS1, KIR412, KKA3, NKAT-8, NKAT8
          CD158E2, KIR-123FM, KIR-G1, NKAT-10, NKAT10
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          Chr.19: 54832676 - 54848569 on Build GRCh38
          Chromosome Location:
          -
          UniGene ID:
          Hs.744319
          Hs.512572
          Hs.654608
          Hs.743496
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID s7853

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KIR2DS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_014512.1 NP_055327.1 6 773
          GenBank mRNA AF022046.1 - 6 773
          AF285437.1 - 5 720
          AM999888.1 - 6 768
          X89892.1 - 6 780
          X98858.1 - 6 760
          Gene Symbol: KIR2DS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291695.1 NP_001278624.1 6 795
          NM_001291696.1 NP_001278625.1 5 495
          NM_001291700.1 NP_001278629.1 4 201
          NM_001291701.1 NP_001278630.1 5 744
          NM_012312.4 NP_036444.1 6 795
          GenBank mRNA AF074087.1 - 4 157
          AF285438.1 - 5 720
          AM409094.1 - 6 761
          AY366243.1 - 6 770
          AY366244.1 - 6 759
          BC108917.1 - 6 762 40017299
          BC119631.1 - 6 779 40115568
          BC119632.1 - 6 779 40115570
          BC125146.1 - 5 744
          BC126856.1 - 6 762 40017301
          L41347.1 - 6 779
          L76667.1 - 6 477
          L76668.1 - 5 471
          U24079.1 - 6 758
          X89893.1 - 6 760
          Gene Symbol: KIR2DS4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001281971.1 NP_001268900.1 6 827
          NM_001281972.1 NP_001268901.1 5 776
          NM_012314.5 NP_036446.3 7 849
          GenBank mRNA AF002255.1 - 7 806
          AJ417554.1 - 5 718
          AJ417555.1 - 5 740
          AJ620615.2 - 5 711
          AJ620616.2 - 5 711
          AY366245.1 - 6 794
          AY366246.1 - 6 745
          AY388472.1 - 6 798
          AY789064.2 - 6 722
          BC096694.1 - 6 771 40026979
          BC101977.1 - 6 771 40004312
          BC103693.1 - 6 771 40004314
          BC108918.1 - 7 762 40017300
          BC125147.1 - 5 744
          BC127084.1 - 6 777 40120468
          BC144566.1 - 6 849 9053094
          BC171788.1 - 6 849 9054442
          L76671.1 - 6 771
          U24077.1 - 6 751
          X94609.1 - 6 815
          Gene Symbol: KIR3DS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001083539.2 NP_001077008.1 7, 8 1082
          NM_001282170.1 NP_001269099.1 6, 7 797
          NM_001282171.1 NP_001269100.1 5, 6 761
          GenBank mRNA AF022044.1 - 7, 8 1058
          AJ417558.3 - 5, 6 994
          AK301140.1 - 7, 8 1082
          BC098107.1 - 5, 6 735 40026977
          BC098288.1 - 7, 8 1056 40026980
          BC105679.1 - 7, 8 1056 40004313
          EF370406.1 - 6, 7 1005
          EF582383.1 - 7, 8 1045
          L76661.1 - 7, 8 1056
          U73396.1 - 7, 8 1045
          X97232.1 - 6, 7 760
          X97233.1 - 7, 8 1045

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          immune response
          signal transduction
          innate immune response
          regulation of immune response
          natural killer cell activation
          transmembrane signaling receptor activity
          MHC class I receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0