Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KIR2DS1 SIRNA Assays ›
          Gene Symbol:
          KIR2DS1 KIR2DL1 KIR2DS5
          Gene Name:
          killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 1
          killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 1
          killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 5
          Gene Aliases:
          CD158H, CD158a, KIR2DP1DL1, KIR2DS4, p50.1
          CD158A, KIR-K64, KIR221, KIR2DS1, NKAT, NKAT-1, NKAT1, p58.1
          CD158G, NKAT9
          Chromosome Location:
          -
          Chromosome Location:
          Chr.19: 54769208 - 54784326 on Build GRCh38
          Chromosome Location:
          -
          UniGene ID:
          Hs.744319
          Hs.654605
          Hs.744266
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s7854

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KIR2DS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_014512.1 NP_055327.1 4 637
          GenBank mRNA AF022046.1 - 4 637
          AF285437.1 - 3 584
          AM999888.1 - 4 632
          X89892.1 - 4 644
          X98858.1 - 4 624
          Gene Symbol: KIR2DL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_014218.2 NP_055033.2 4 662
          XM_011526939.2 XP_011525241.1 4 1267
          XM_017026782.1 XP_016882271.1 4 640
          XM_017026783.1 XP_016882272.1 4 1267
          GenBank mRNA AF022045.1 - 4 631
          AF135560.1 - 2 561
          AF285431.1 - 4 657
          AF285432.1 - 3 584
          AY366237.1 - 4 660
          AY366238.1 - 4 606
          AY366239.1 - 4 654
          AY366240.1 - 4 662
          AY366241.1 - 4 661
          AY388471.1 - 4 654
          AY789055.1 - 5 657
          AY789056.1 - 4 624
          BC069344.1 - 4 657 7262296
          KJ699235.1 - 4 897
          L41267.1 - 5 656
          U24076.1 - 5 657
          U24078.1 - 4 664
          U96189.1 - 2 561
          Gene Symbol: KIR2DS5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_014513.2 NP_055328.2 4 658
          GenBank mRNA AF208054.1 - 4 658
          AF272389.1 - 5 608
          AY366247.1 - 4 682
          AY366248.1 - 6 682
          L76672.1 - 4 635

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          immune response
          signal transduction
          innate immune response
          regulation of immune response
          natural killer cell inhibitory signaling pathway
          transmembrane signaling receptor activity
          receptor activity
          protein binding
          HLA-C specific inhibitory MHC class I receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0