Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPLP0 SIRNA Assays ›
          Gene Symbol:
          RPLP0 RPLP0P2 TNS1
          Gene Name:
          ribosomal protein lateral stalk subunit P0
          ribosomal protein lateral stalk subunit P0 pseudogene 2
          tensin 1
          Gene Aliases:
          L10E, LP0, P0, PRLP0, RPP0
          RPLP0L2, RPLP0_3_1146
          MST091, MST122, MST127, MSTP091, MSTP122, MSTP127, MXRA6, PPP1R155, TNS
          Chromosome Location:
          Chr.12: 120196700 - 120201211 on Build GRCh38
          Chromosome Location:
          Chr.11: 61615036 - 61639449 on Build GRCh38
          Chromosome Location:
          Chr.2: 217799789 - 217978853 on Build GRCh38
          UniGene ID:
          Hs.546285
          Hs.502733
          Hs.471381
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s786
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RPLP0
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001002.3 NP_000993.1 8 1128
          NM_053275.3 NP_444505.1 8 1188
          GenBank mRNA AF274958.1 - 1 441
          AK001313.1 - 6 2524
          AK129754.1 - 8 1024
          AK129823.1 - 8 1027
          AK222468.1 - 1 1026
          AK222571.1 - 8 1024
          AK291109.1 - 8 1026
          AK304674.1 - 8 871
          BC000087.2 - 1 1012 3507437
          BC000345.2 - 1 1000 2960918
          BC000752.2 - 1 1000 2964581
          BC001127.2 - 1 993 2987976
          BC001834.2 - 1 1000 2964581
          BC003655.2 - 7 1058 3541947
          BC005863.2 - 1 1000 2960918
          BC008092.1 - 7 1004 3458803
          BC008594.1 - 1 995 3463159
          BC009867.2 - 1 1006 3940316
          BC015173.1 - 7 1000 3903073
          BC015690.1 - 1 1014 4660892
          BC019014.2 - 7 1069
          BC070194.1 - 2 833 6457312
          BC104645.1 - 1 472
          BC107717.1 - 2 842 6736210
          M17885.1 - 8 1028
          Gene Symbol: RPLP0P2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002775.2 5 1713
          GenBank mRNA AK095332.1 - 5 1713
          Gene Symbol: TNS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK309785.1 - 2528

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs99999902_m1

          Panther Classification:

          Molecular Function -

          ribosomal protein translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
          cytoplasmic translation
          rRNA processing
          translation
          translational initiation
          SRP-dependent cotranslational protein targeting to membrane
          viral transcription
          structural constituent of ribosome
          protein binding
          poly(A) RNA binding
          large ribosomal subunit rRNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0