Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ech1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016772.1 | NP_058052.1 | 2 | 181 | ||
GenBank mRNA | AF030343.1 | - | 2 | 181 | ||
AK088495.1 | - | 2 | 169 | |||
BC068112.1 | - | 2 | 181 | 5710971 | ||
BC087924.1 | - | 2 | 242 | 6706361 |