Search
Search

Wwc2| Gene Symbol: Wwc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133791.4 | NP_598552.2 | 17, 18 | 3047 | ||
| GenBank mRNA | AB045323.1 | - | 17, 18 | 3072 | ||
| AK028956.1 | - | 664 | ||||
| AK032989.1 | - | 13, 14 | 2113 | |||
| AK079375.1 | - | 17, 18 | 2883 | |||
| AK147648.1 | - | 3041 | ||||
| BC054434.1 | - | 7, 8 | 1328 | |||
| BC067050.1 | - | 17, 18 | 3047 | 30537299 | ||