Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024499.1 | NP_078775.1 | 4, 5 | 352 | ||
GenBank mRNA | AK039966.1 | - | 3, 4 | 347 | ||
AK051005.1 | - | 4, 5 | 351 | |||
AK079168.1 | - | 4, 5 | 362 | |||
AK165557.1 | - | 4, 5 | 376 | |||
AK189623.1 | - | 1, 2 | 61 | |||
AK193443.1 | - | 1, 2 | 63 | |||
BC003836.1 | - | 4, 5 | 352 | 3486148 |