Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016899.4 | NP_058595.2 | 4, 5 | 712 | ||
GenBank mRNA | AB232620.1 | - | 4, 5 | 508 | ||
AF119675.1 | - | 4, 5 | 648 | |||
AK003260.1 | - | 4, 5 | 713 | |||
BC006624.1 | - | 4, 5 | 698 | 3968810 |