Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017397.3 | NP_059093.3 | 5 | 959 | ||
GenBank mRNA | AF217000.1 | - | 5 | 834 | ||
AF217001.1 | - | 5 | 834 | |||
AF220454.1 | - | 5 | 962 | |||
AJ250027.1 | - | 5 | 834 | |||
AK050950.1 | - | 5 | 833 | |||
AK154518.1 | - | 5 | 838 | |||
BC125471.1 | - | 5 | 959 | 40129986 | ||
BC137721.1 | - | 5 | 959 | 8860740 |