Search
Search

Cks1b| Gene Symbol: Cks1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016904.1 | NP_058600.1 | 1 | 122 | ||
| GenBank mRNA | AB025409.1 | - | 1 | 122 | ||
| AK010414.2 | - | 1 | 109 | |||
| AK011782.1 | - | 1 | 88 | |||
| BC076579.1 | - | 1 | 114 | 6846517 | ||