Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KRT8 SIRNA Assays ›
          Gene Symbol:
          KRT8 KRT8P35 KRT8P41
          Gene Name:
          keratin 8
          keratin 8 pseudogene 35
          keratin 8 pseudogene 41
          Gene Aliases:
          CARD2, CK-8, CK8, CYK8, K2C8, K8, KO
          -
          -
          Chromosome Location:
          Chr.12: 52897187 - 52949866 on Build GRCh38
          Chromosome Location:
          Chr.3: 63096246 - 63097755 on Build GRCh38
          Chromosome Location:
          Chr.11: 9094363 - 9096190 on Build GRCh38
          UniGene ID:
          Hs.533782
          Hs.590045
          Hs.689472
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s7968

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KRT8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256282.1 NP_001243211.1 4 653
          NM_001256293.1 NP_001243222.1 4 747
          NM_002273.3 NP_002264.1 3 652
          NR_045962.1 4 1006
          GenBank mRNA AK222941.1 - 3 625
          AK290938.1 - 4 705
          AK310257.1 - 4 653
          AK315826.1 - 4 982
          BC000654.2 - 3 613 3349233
          BC008200.1 - 3 267 3445666
          BC011373.1 - 2 195
          BC063513.1 - 3 617 3879823
          BC073760.1 - 3 599 2963092
          BC075839.1 - 3 597 6285632
          BX417476.2 - 3 639
          M26512.1 - 3 584
          M34225.1 - 3 583
          M77025.1 - 3 262
          U76549.1 - 3 565
          X74929.1 - 3 608
          Gene Symbol: KRT8P35
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK091178.1 - 1 809
          Gene Symbol: KRT8P41
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027713.1 1 667
          GenBank mRNA AK127993.1 - 1 667

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          viral process
          tumor necrosis factor-mediated signaling pathway
          sarcomere organization
          response to hydrostatic pressure
          response to other organism
          cell differentiation involved in embryonic placenta development
          extrinsic apoptotic signaling pathway
          hepatocyte apoptotic process
          structural molecule activity
          protein binding
          protein complex binding
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8qtqx:80/100.66.78.82:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0