Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dfna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018769.3 | NP_061239.1 | 3, 4 | 465 | ||
GenBank mRNA | AF073309.1 | - | 3, 4 | 466 | ||
AK016561.1 | - | 833 | ||||
AK089459.1 | - | 452 | ||||
AK155135.1 | - | 444 | ||||
AK171278.1 | - | 3, 4 | 464 | |||
BC051962.1 | - | 3, 4 | 525 | |||
BC086618.1 | - | 3, 4 | 525 | |||
BC132303.1 | - | 2, 3 | 436 | 40130580 |