Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018888.4 | NP_061376.2 | 7, 8 | 844 | ||
GenBank mRNA | AF253516.1 | - | 7, 8 | 591 | ||
AK010376.1 | - | 7, 8 | 585 | |||
AK011152.1 | - | 7, 8 | 598 | |||
AK011953.1 | - | 7, 8 | 568 | |||
AK077323.1 | - | 7, 8 | 582 | |||
AK139488.1 | - | 7, 8 | 844 | |||
AK169413.1 | - | 7, 8 | 581 | |||
BC050884.1 | - | 7, 8 | 797 | 6808756 | ||
BC057570.1 | - | 7, 8 | 797 | 6808756 |