Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GSK3B SIRNA Assays ›
          Gene Symbol:
          Gsk3b
          Gene Name:
          glycogen synthase kinase 3 beta
          Gene Aliases:
          7330414F15Rik, 8430431H08Rik, C86142, GSK-3, GSK-3beta, GSK3
          Chromosome Location:
          Chr.16: 38089001 - 38246080 on Build GRCm38
          UniGene ID:
          Mm.394930
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s80827

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gsk3b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019827.6 NP_062801.1 9, 10 2624
          GenBank mRNA AB066114.1 - 10, 11 2623
          AF156099.2 - 9, 10 1265
          AK154293.1 - 10, 11 2102
          AK170965.1 - 10, 11 2071
          BC006936.1 - 9, 10 1131 2648507
          BC060743.1 - 10, 11 2551 4022374

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001146156, NM_002093, AK290897, L33801, CR536510, and others.
          This siRNA also targets the following transcripts in Rat: NM_032080, XM_006248375, XM_008768745, XM_006248373, XM_006248374, and others.

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          re-entry into mitotic cell cycle
          epithelial to mesenchymal transition
          positive regulation of cell-matrix adhesion
          carbohydrate metabolic process
          glycogen metabolic process
          regulation of gene expression by genetic imprinting
          protein phosphorylation
          protein export from nucleus
          ER overload response
          cytoskeleton organization
          establishment or maintenance of cell polarity
          multicellular organism development
          nervous system development
          axonogenesis
          myoblast fusion
          circadian rhythm
          cell proliferation
          organ morphogenesis
          negative regulation of signal transduction
          negative regulation of cardiac muscle hypertrophy
          positive regulation of peptidyl-threonine phosphorylation
          regulation of neuron projection development
          negative regulation of neuron projection development
          negative regulation of neuron maturation
          myotube differentiation
          Wnt signaling pathway
          phosphorylation
          cell migration
          peptidyl-serine phosphorylation
          hippocampus development
          establishment of cell polarity
          cell differentiation
          negative regulation of protein complex assembly
          positive regulation of protein complex assembly
          negative regulation of protein binding
          positive regulation of protein binding
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          regulation of microtubule-based process
          positive regulation of peptidyl-serine phosphorylation
          protein localization to microtubule
          intracellular signal transduction
          cellular response to interleukin-3
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of MAP kinase activity
          positive regulation of GTPase activity
          hypermethylation of CpG island
          canonical Wnt signaling pathway involved in positive regulation of apoptotic process
          fat cell differentiation
          positive regulation of axon extension
          positive regulation of transcription from RNA polymerase II promoter
          protein autophosphorylation
          positive regulation of protein export from nucleus
          regulation of neuronal synaptic plasticity
          rhythmic process
          negative regulation of dendrite morphogenesis
          negative regulation of NFAT protein import into nucleus
          canonical Wnt signaling pathway
          superior temporal gyrus development
          cellular response to mechanical stimulus
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
          positive regulation of stem cell differentiation
          nucleotide binding
          RNA polymerase II transcription factor binding
          p53 binding
          protein kinase activity
          protein serine/threonine kinase activity
          integrin binding
          protein binding
          ATP binding
          beta-catenin binding
          transcription factor binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein kinase binding
          ubiquitin protein ligase binding
          protein kinase A catalytic subunit binding
          ionotropic glutamate receptor binding
          tau protein binding
          tau-protein kinase activity
          NF-kappaB binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0