Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LCAT SIRNA Assays ›
          Gene Symbol:
          LCAT SLC12A4
          Gene Name:
          lecithin-cholesterol acyltransferase
          solute carrier family 12 member 4
          Gene Aliases:
          -
          CTC-479C5.17, KCC1, hKCC1
          Chromosome Location:
          Chr.16: 67939884 - 67944112 on Build GRCh38
          Chromosome Location:
          Chr.16: 67943474 - 67968694 on Build GRCh38
          UniGene ID:
          Hs.387239
          Hs.10094
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s8103

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LCAT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000229.1 NP_000220.1 1 137
          GenBank mRNA BC014781.1 - 1 127 4155094
          BT009748.1 - 1 126
          M12625.1 - 1 393
          M26268.1 - 1 79
          X06537.1 - 1 229
          Gene Symbol: SLC12A4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145961.1 NP_001139433.1 24 4245
          NM_001145962.1 NP_001139434.1 23 4168
          NM_001145963.1 NP_001139435.1 24 4142
          NM_001145964.1 NP_001139436.1 24 4133
          NM_005072.4 NP_005063.1 24 4263
          GenBank mRNA AK310027.1 - 1 545

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          acyltransferase secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          phospholipid metabolic process
          phosphatidylcholine biosynthetic process
          cholesterol metabolic process
          cholesterol transport
          very-low-density lipoprotein particle remodeling
          high-density lipoprotein particle remodeling
          cholesterol esterification
          lipoprotein metabolic process
          lipoprotein biosynthetic process
          cholesterol homeostasis
          reverse cholesterol transport
          phosphatidylcholine metabolic process
          response to copper ion
          response to glucocorticoid
          regulation of high-density lipoprotein particle assembly
          transport
          ion transport
          cell volume homeostasis
          chemical synaptic transmission
          potassium ion transmembrane transport
          chloride transmembrane transport
          phosphatidylcholine-sterol O-acyltransferase activity
          phospholipase A2 activity
          protein binding
          apolipoprotein A-I binding
          potassium:chloride symporter activity
          protein kinase binding
          potassium ion symporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-gbx7x:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0