Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205085.1 | NP_001192014.1 | 5 | 1259 | ||
NM_020496.3 | NP_065242.1 | 5 | 1259 | |||
NM_194263.2 | NP_919239.1 | 5 | 1259 | |||
GenBank mRNA | AF260557.1 | - | 5 | 1264 | ||
AF306667.1 | - | 5 | 1197 | |||
AJ277486.1 | - | 5 | 801 | |||
AK052155.1 | - | 5 | 1241 | |||
AK052389.1 | - | 5 | 1242 | |||
AK141251.1 | - | 5 | 1029 | |||
BY385297.1 | - | 3 | 290 |