siRNA Details
Gene Symbol: Gabrq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290435.1 | NP_001277364.1 | 3 | 564 | ||
NM_020488.2 | NP_065234.1 | 3 | 564 | |||
GenBank mRNA | AF189260.1 | - | 3 | 306 | ||
AK038859.1 | - | 3 | 475 | |||
AK156674.1 | - | 3 | 565 | |||
BC119074.1 | - | 3 | 304 | 8733823 | ||
BC119076.1 | - | 3 | 304 | 8733825 |

More Information
Other Species Targeted

Panther Classification:
Molecular Function -






