Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020566.1 | NP_065591.1 | 5 | 584 | ||
GenBank mRNA | AF036875.1 | - | 5 | 584 | ||
AK008507.1 | - | 5 | 833 | |||
AK170008.1 | - | 5 | 826 | |||
BC002305.1 | - | 2 | 218 | |||
BC049281.1 | - | 5 | 818 | 4989485 |