Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163753.1 | NP_001157225.1 | 4 | 702 | ||
NM_021411.4 | NP_067386.3 | 4 | 314 | |||
GenBank mRNA | AB232633.1 | - | 4 | 293 | ||
AF233582.1 | - | 4 | 293 | |||
AK039716.1 | - | 4 | 308 | |||
AK040216.1 | - | 4 | 314 | |||
AK045995.1 | - | 4 | 305 | |||
AK046260.1 | - | 4 | 703 | |||
AK050483.1 | - | 4 | 308 | |||
BC050757.1 | - | 4 | 314 | |||
BC128020.1 | - | 4 | 312 | 40136290 |