Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LHX1 SIRNA Assays ›
          Gene Symbol:
          LHX1
          Gene Name:
          LIM homeobox 1
          Gene Aliases:
          LIM-1, LIM1
          Chromosome Location:
          Chr.17: 36937475 - 36944615 on Build GRCh38
          UniGene ID:
          Hs.443727
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s8166

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LHX1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005568.4 NP_005559.2 1, 2 899
          GenBank mRNA AK298253.1 - 1, 2 542
          BC020470.1 - 1, 2 899
          BC101674.1 - 1, 2 268 8069180
          BC111936.1 - 1, 2 268 8327404
          U14755.1 - 1, 2 608

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          helix-turn-helix transcription factor homeodomain transcription factor DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          urogenital system development
          ureteric bud development
          branching involved in ureteric bud morphogenesis
          gastrulation with mouth forming second
          ectoderm formation
          endoderm formation
          kidney development
          transcription from RNA polymerase II promoter
          cell-cell signaling
          pattern specification process
          nervous system development
          motor neuron axon guidance
          anatomical structure morphogenesis
          post-embryonic development
          embryonic pattern specification
          organ morphogenesis
          anterior/posterior axis specification
          anterior/posterior pattern specification
          dorsal/ventral pattern formation
          regulation of gene expression
          retina layer formation
          ventral spinal cord development
          spinal cord association neuron differentiation
          telencephalon development
          cerebellum development
          cerebellar Purkinje cell differentiation
          forebrain regionalization
          cerebellar Purkinje cell-granule cell precursor cell signaling involved in regulation of granule cell precursor cell proliferation
          somite rostral/caudal axis specification
          metanephric part of ureteric bud development
          oviduct epithelium development
          uterine epithelium development
          nephric duct elongation
          horizontal cell localization
          positive regulation of embryonic development
          cellular response to fibroblast growth factor stimulus
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          anatomical structure formation involved in morphogenesis
          embryonic viscerocranium morphogenesis
          pronephros development
          embryonic retina morphogenesis in camera-type eye
          uterus development
          oviduct development
          cervix development
          vagina development
          head development
          epithelium development
          paramesonephric duct development
          comma-shaped body morphogenesis
          S-shaped body morphogenesis
          renal vesicle morphogenesis
          mesonephric duct development
          nephric duct morphogenesis
          metanephric glomerulus development
          metanephric comma-shaped body morphogenesis
          metanephric renal vesicle morphogenesis
          metanephric S-shaped body morphogenesis
          primitive streak formation
          positive regulation of branching involved in ureteric bud morphogenesis
          lateral motor column neuron migration
          positive regulation of gastrulation
          positive regulation of anterior head development
          positive regulation of nephron tubule epithelial cell differentiation
          transcription factor activity, sequence-specific DNA binding
          transcription corepressor activity
          zinc ion binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0