Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021500.2 | NP_067475.2 | 8 | 964 | ||
GenBank mRNA | AF263247.1 | - | 8 | 926 | ||
AK034859.1 | - | 8 | 963 | |||
AK049424.1 | - | 8 | 984 | |||
AK075637.1 | - | 7 | 804 | |||
AK088854.1 | - | 8 | 972 | |||
AK145800.1 | - | 8 | 1027 | |||
AK152523.1 | - | 8 | 972 | |||
AK168788.1 | - | 8 | 990 | |||
AK169149.1 | - | 8 | 972 | |||
BC039054.1 | - | 8 | 950 | 2648667 | ||
BC058687.1 | - | 8 | 962 | 6411707 |