Search
Search

Pcbp4| Gene Symbol: Pcbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021567.5 | NP_067542.3 | 9 | 849 | ||
| GenBank mRNA | AF176328.1 | - | 9 | 735 | ||
| AK004580.1 | - | 9 | 800 | |||
| AK005383.1 | - | 9 | 798 | |||
| AK077632.1 | - | 9 | 799 | |||
| AK082005.1 | - | 9 | 799 | |||
| BC010694.1 | - | 9 | 738 | 2654291 | ||