Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021713.2 | NP_068359.1 | 2, 3 | 361 | ||
GenBank mRNA | AF289484.1 | - | 2, 3 | 353 | ||
AK002656.1 | - | 2, 3 | 336 | |||
AK004104.1 | - | 2, 3 | 336 | |||
AK013232.1 | - | 2, 3 | 307 | |||
BC028501.1 | - | 2, 3 | 346 | 5256915 | ||
CJ180315.1 | - | 2, 3 | 361 |