Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110807.1 | NP_001104277.1 | 2 | 332 | ||
GenBank mRNA | AJ289243.1 | - | 2 | 340 | ||
BC028751.1 | - | 2 | 331 | 1314413 |