Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021899.3 | NP_068699.1 | 2, 3 | 1133 | ||
GenBank mRNA | AB221666.1 | - | 1, 2 | 332 | ||
AF253052.1 | - | 2, 3 | 1145 | |||
AK154574.1 | - | 2, 3 | 1376 | |||
BC040395.1 | - | 3, 4 | 763 | 4190087 |