Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022016.3 | NP_071299.3 | 14 | 2010 | ||
GenBank mRNA | AF229929.1 | - | 14 | 1921 | ||
AF266478.1 | - | 14 | 2056 | |||
BC022970.1 | - | 14 | 1998 | 5366008 |