Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146217.1 | NP_001139689.1 | 10 | 1246 | ||
NM_022316.2 | NP_071711.2 | 10 | 1213 | |||
GenBank mRNA | AF070470.1 | - | 10 | 1189 | ||
AK040931.1 | - | 10 | 1247 | |||
BC031804.1 | - | 10 | 1193 | 3673732 |