Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122765.1 | NP_001116237.1 | 12 | 907 | ||
NM_001122766.1 | NP_001116238.1 | 10 | 744 | |||
NM_022432.4 | NP_071877.3 | 13 | 954 | |||
GenBank mRNA | AB221570.1 | - | 13 | 859 | ||
AF299337.1 | - | 13 | 939 | |||
AK014042.1 | - | 12 | 878 | |||
AK144431.1 | - | 12 | 851 | |||
AK146412.1 | - | 12 | 884 | |||
AK183534.1 | - | 5 | 328 | |||
BC021439.1 | - | 13 | 958 | 5039278 |