Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LMO7 SIRNA Assays ›
Gene Symbol:
LMO7
Gene Name:
LIM domain 7
Gene Aliases:
FBX20, FBXO20, LOMP
Chromosome Location:
Chr.13: 75620434 - 75859870 on Build GRCh38
UniGene ID:
Hs.207631
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s8237
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LMO7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306080.1
NP_001293009.1
27
4612
NM_005358.5
NP_005349.3
26
4656
NM_015842.2
NP_056667.2
24
4303
XM_011535075.2
XP_011533377.1
28
4500
XM_011535078.1
XP_011533380.1
27
4458
XM_011535080.2
XP_011533382.1
28
4419
XM_011535082.1
XP_011533384.1
27
4377
XM_011535086.1
XP_011533388.1
26
4173
XM_011535094.1
XP_011533396.1
21
3081
XM_011535095.1
XP_011533397.1
24
3252
XM_017020574.1
XP_016876063.1
28
4500
XM_017020575.1
XP_016876064.1
27
4458
XM_017020576.1
XP_016876065.1
27
4443
XM_017020577.1
XP_016876066.1
28
4419
XM_017020578.1
XP_016876067.1
26
4401
XM_017020579.1
XP_016876068.1
27
4377
XM_017020580.1
XP_016876069.1
27
4296
XM_017020581.1
XP_016876070.1
27
4422
XM_017020582.1
XP_016876071.1
27
4215
XM_017020583.1
XP_016876072.1
27
4215
XM_017020584.1
XP_016876073.1
26
4218
XM_017020585.1
XP_016876074.1
26
4014
XM_017020586.1
XP_016876075.1
26
3933
XM_017020587.1
XP_016876076.1
25
3891
XM_017020588.1
XP_016876077.1
25
3972
XM_017020589.1
XP_016876078.1
25
4095
XM_017020590.1
XP_016876079.1
25
3891
XM_017020591.1
XP_016876080.1
27
4583
XM_017020592.1
XP_016876081.1
27
4466
XM_017020593.1
XP_016876082.1
26
4379
XM_017020594.1
XP_016876083.1
26
4262
XM_017020595.1
XP_016876084.1
25
4256
XM_017020596.1
XP_016876085.1
25
3294
XM_017020597.1
XP_016876086.1
24
3264
XM_017020598.1
XP_016876087.1
25
4256
XM_017020599.1
XP_016876088.1
25
4097
XM_017020600.1
XP_016876089.1
25
3213
XM_017020601.1
XP_016876090.1
24
3974
XM_017020602.1
XP_016876091.1
24
3857
XM_017020603.1
XP_016876092.1
24
3974
XM_017020604.1
XP_016876093.1
24
3377
GenBank mRNA
AB020665.2
-
24
4233
AF144237.1
-
15
2116
AF330045.1
-
26
4655
AK074265.1
-
11
1106
AK304051.1
-
22
3194
FJ711162.1
-
27
4242
U90654.1
-
4
372
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein ubiquitination
regulation of signaling
regulation of cell adhesion
ubiquitin-protein transferase activity
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-gk48b:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0