Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023146.2 | NP_075635.1 | 12 | 1489 | ||
GenBank mRNA | AJ271458.1 | - | 12 | 1387 | ||
AK016583.1 | - | 12 | 1389 | |||
AK035897.1 | - | 12 | 1386 | |||
AK046865.1 | - | 12 | 1375 | |||
AK050150.1 | - | 8 | 940 | |||
AK139704.1 | - | 12 | 1397 | |||
AK163961.1 | - | 8 | 895 | |||
BC033503.1 | - | 12 | 1383 |