Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025430.3 | NP_079706.1 | 3 | 303 | ||
GenBank mRNA | AK004385.1 | - | 3 | 296 | ||
AK010223.1 | - | 3 | 292 | |||
AK010369.1 | - | 3 | 289 | |||
AK012807.1 | - | 3 | 290 | |||
AK039187.1 | - | 3 | 283 | |||
BB615775.1 | - | 3 | 303 | |||
BC028750.1 | - | 3 | 295 | 1245736 |