Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024172.3 | NP_077134.1 | 5 | 847 | ||
GenBank mRNA | AF338351.1 | - | 4 | 785 | ||
AK013263.1 | - | 5 | 838 | |||
AK088469.1 | - | 5 | 823 | |||
AK160128.1 | - | 5 | 842 | |||
BC014758.1 | - | 5 | 847 | 4014661 |