Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290706.1 | NP_001277635.1 | 3 | 409 | ||
NM_025452.4 | NP_079728.2 | 3 | 427 | |||
GenBank mRNA | AK008935.1 | - | 3 | 342 | ||
BC019563.1 | - | 3 | 356 | 4219997 | ||
BY079646.1 | - | 3 | 375 |