siRNA Details
Gene Symbol: Tmem54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290706.1 | NP_001277635.1 | 4, 5 | 623 | ||
NM_025452.4 | NP_079728.2 | 4, 5 | 641 | |||
NM_133881.1 | NP_598642.1 | 3, 4 | 581 | |||
GenBank mRNA | AK007525.1 | - | 3, 4 | 497 | ||
AK008935.1 | - | 4, 5 | 556 | |||
AK009830.1 | - | 3, 4 | 477 | |||
BC019563.1 | - | 4, 5 | 570 | 4219997 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
