Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163536.1 | NP_001157008.1 | 2 | 170 | ||
NM_001163537.1 | NP_001157009.1 | 2 | 190 | |||
NM_025489.3 | NP_079765.3 | 2 | 177 | |||
GenBank mRNA | AK005725.1 | - | 2 | 157 | ||
AK005749.1 | - | 2 | 146 | |||
BC049579.1 | - | 2 | 120 | 6772915 | ||
BY085815.1 | - | 2 | 171 |