Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LTBP1 SIRNA Assays ›
Gene Symbol:
LTBP1
Gene Name:
latent transforming growth factor beta binding protein 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 32947129 - 33399509 on Build GRCh38
UniGene ID:
Hs.619315
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s8317
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LTBP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000627.3
NP_000618.3
29
4105
NM_001166264.1
NP_001159736.1
28
3979
NM_001166265.1
NP_001159737.1
29
3946
NM_001166266.1
NP_001159738.1
28
3820
NM_206943.2
NP_996826.2
33
4965
XM_005264317.3
XP_005264374.1
33
4808
XM_005264318.3
XP_005264375.1
32
4682
XM_011532853.2
XP_011531155.1
33
5141
XM_011532855.2
XP_011531157.1
33
4964
XM_011532856.2
XP_011531158.1
32
4850
XM_011532857.2
XP_011531159.1
32
4844
XM_011532858.2
XP_011531160.1
32
4844
XM_011532859.2
XP_011531161.1
33
4811
XM_011532860.2
XP_011531162.1
32
4802
XM_011532861.2
XP_011531163.1
31
4700
XM_017004108.1
XP_016859597.1
32
4847
XM_017004109.1
XP_016859598.1
32
4841
XM_017004110.1
XP_016859599.1
31
4721
GenBank mRNA
AB208801.1
-
29
3904
AK298027.1
-
23
2674
AK299237.1
-
11
1450
AK299912.1
-
23
2777
AK302857.1
-
14
1844
AK315991.1
-
14
2008
BC130289.1
-
29
4045
40146320
BC144127.1
-
28
3762
BC144128.1
-
28
3919
9052648
BX103725.1
-
1
107
Back To Top
More Information
Panther Classification:
Molecular Function -
extracellular matrix structural protein
Gene Ontology Categories:
Function(s)
Process(es)
ventricular septum development
transmembrane receptor protein serine/threonine kinase signaling pathway
sequestering of TGFbeta in extracellular matrix
aorta development
coronary vasculature development
transforming growth factor beta-activated receptor activity
calcium ion binding
protein binding
transforming growth factor beta binding
microfibril binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0