Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LTF SIRNA Assays ›
          Gene Symbol:
          LTF
          Gene Name:
          lactotransferrin
          Gene Aliases:
          GIG12, HEL110, HLF2, LF
          Chromosome Location:
          Chr.3: 46436005 - 46485234 on Build GRCh38
          UniGene ID:
          Hs.529517
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s8330

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LTF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199149.1 NP_001186078.1 2 349
          NM_001321121.1 NP_001308050.1 2 460
          NM_001321122.1 NP_001308051.1 5 693
          NM_002343.5 NP_002334.2 2 460
          GenBank mRNA AF332168.1 - 2 167
          AK093852.1 - 5 696
          AK290859.1 - 2 205
          AK292813.1 - 2 205
          AK298035.1 - 2 349
          AK303889.1 - 2 205
          AK316330.1 - 2 269
          AY137470.1 - 2 167
          AY165046.1 - 2 179
          AY178998.2 - 2 167
          AY360320.1 - 2 168
          AY493417.1 - 2 277
          AY875691.1 - 2 167
          BC015822.1 - 2 209 4251222
          BC015823.1 - 2 211 4294752
          BC022347.1 - 2 205 4251058
          BP342557.1 - 2 408
          CD722125.1 - 2 193
          DQ522304.1 - 2 167
          EU779935.1 - 2 167
          EU794669.1 - 2 202
          KF992208.1 - 2 179
          KT006756.1 - 2 167
          M83202.1 - 2 174
          M83205.1 - 2 153
          M93150.1 - 2 184
          U07643.1 - 2 264
          X52941.1 - 2 161
          X53961.1 - 2 461

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          regulation of cytokine production
          retina homeostasis
          innate immune response in mucosa
          transcription, DNA-templated
          proteolysis
          ion transport
          humoral immune response
          antibacterial humoral response
          antifungal humoral response
          negative regulation of lipopolysaccharide-mediated signaling pathway
          regulation of tumor necrosis factor production
          negative regulation of ATPase activity
          iron assimilation by chelation and transport
          positive regulation of osteoblast proliferation
          positive regulation of toll-like receptor 4 signaling pathway
          negative regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          cellular protein metabolic process
          negative regulation by host of viral process
          negative regulation of viral genome replication
          cell redox homeostasis
          positive regulation of osteoblast differentiation
          negative regulation of viral process
          positive regulation of NF-kappaB transcription factor activity
          bone morphogenesis
          positive regulation of protein serine/threonine kinase activity
          positive regulation of bone mineralization involved in bone maturation
          negative regulation of single-species biofilm formation in or on host organism
          positive regulation of chondrocyte proliferation
          negative regulation of tumor necrosis factor (ligand) superfamily member 11 production
          negative regulation of osteoclast development
          DNA binding
          serine-type endopeptidase activity
          iron ion binding
          protein binding
          heparin binding
          protein serine/threonine kinase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-zxk44:80/100.66.78.82:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0