Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025576.2 | NP_079852.1 | 1, 2 | 449 | ||
GenBank mRNA | AK012674.1 | - | 1, 2 | 439 | ||
AK014691.1 | - | 1, 2 | 443 | |||
AK151972.1 | - | 1, 2 | 450 | |||
AK168416.1 | - | 257 | ||||
BC026750.1 | - | 1, 2 | 428 | 5135841 |