Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025624.2 | NP_079900.1 | 4 | 243 | ||
GenBank mRNA | AK003439.1 | - | 4 | 236 | ||
AK011115.1 | - | 4 | 244 | |||
AK012455.1 | - | 4 | 205 | |||
AK013934.1 | - | 4 | 237 | |||
AK014296.1 | - | 4 | 207 | |||
AK044283.1 | - | 4 | 219 | |||
AK132041.1 | - | 220 | ||||
AK167958.1 | - | 4 | 244 | |||
BC027016.1 | - | 4 | 234 | 5375452 |