Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023229.2 | NP_075718.2 | 5 | 1100 | ||
GenBank mRNA | AF168682.1 | - | 4 | 995 | ||
AK002552.1 | - | 5 | 1049 | |||
AK003940.1 | - | 2 | 343 | |||
AK038596.1 | - | 5 | 1052 | |||
AK052667.1 | - | 5 | 1045 | |||
AK139374.1 | - | 4 | 1819 | |||
AK151859.1 | - | 5 | 1052 | |||
AK152369.1 | - | 5 | 1101 | |||
AK159751.1 | - | 5 | 1052 | |||
AK164317.1 | - | 1058 | ||||
AK166508.1 | - | 1645 | ||||
AK167346.1 | - | 4 | 1812 | |||
AK179538.1 | - | 2 | 269 | |||
BC013547.1 | - | 5 | 1017 | 4218374 |