Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ormdl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025661.4 | NP_079937.1 | 4 | 513 | ||
GenBank mRNA | AK009621.1 | - | 4 | 514 | ||
AK012718.1 | - | 4 | 483 | |||
AK087497.1 | - | 4 | 482 | |||
AK154085.1 | - | 4 | 505 | |||
BC026412.1 | - | 2 | 188 | |||
BC046594.1 | - | 4 | 451 | 6308578 |