Search
Search

SH2D1A| Gene Symbol: SH2D1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114937.2 | NP_001108409.1 | 1, 2 | 507 | ||
| NM_002351.4 | NP_002342.1 | 1, 2 | 507 | |||
| GenBank mRNA | AB586694.1 | - | 1, 2 | 248 | ||
| AF072930.1 | - | 1, 2 | 232 | |||
| AF073019.1 | - | 1, 2 | 232 | |||
| AF100539.1 | - | 1, 2 | 200 | |||
| AF100541.1 | - | 1, 2 | 200 | |||
| AK311911.1 | - | 1, 2 | 181 | |||
| AL023657.1 | - | 1, 2 | 445 | |||
| BC020732.1 | - | 1, 2 | 239 | 4702653 | ||
| CR542031.1 | - | 1, 2 | 146 | |||
| CR542043.1 | - | 1, 2 | 146 | |||
| DA007747.1 | - | 1, 2 | 507 | |||
| JX512648.1 | - | 1, 2 | 179 | |||