Search
Search

Tpgs2| Gene Symbol: Tpgs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004361.2 | NP_001004361.2 | 4, 5 | 640 | ||
| NM_001142697.1 | NP_001136169.1 | 4, 5 | 386 | |||
| NM_001142698.1 | NP_001136170.1 | 4, 5 | 640 | |||
| GenBank mRNA | AK017728.1 | - | 4, 5 | 394 | ||
| AK141632.1 | - | 4, 5 | 458 | |||
| AK160220.1 | - | 4, 5 | 436 | |||
| BC060253.1 | - | 2, 3 | 192 | |||
| BC080777.1 | - | 4, 5 | 396 | 30439612 | ||
| BQ770487.1 | - | 4, 5 | 640 | |||
| DV655125.1 | - | 4, 5 | 386 | |||