Search
Search

Rras2| Gene Symbol: Rras2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025846.2 | NP_080122.2 | 2 | 420 | ||
| GenBank mRNA | AK011419.1 | - | 2 | 420 | ||
| AK078870.1 | - | 2 | 421 | |||
| AK172037.1 | - | 2 | 412 | |||
| AK172113.1 | - | 2 | 406 | |||
| BC003871.1 | - | 2 | 371 | 3492159 | ||