Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080554.2 | NP_542121.2 | 5 | 700 | ||
GenBank mRNA | AK005335.1 | - | 5 | 678 | ||
AK011026.1 | - | 5 | 677 | |||
AK029067.1 | - | 4 | 997 | |||
AK032992.1 | - | 5 | 678 | |||
AK078419.1 | - | 5 | 678 | |||
AK145587.1 | - | 5 | 677 | |||
AK148284.1 | - | 701 | ||||
AK220321.1 | - | 5 | 641 | |||
BC019112.1 | - | 5 | 711 |