Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuedc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164290.1 | NP_001157762.1 | 10 | 976 | ||
NM_001164291.1 | NP_001157763.1 | 9 | 868 | |||
NM_001164292.1 | NP_001157764.1 | 9 | 865 | |||
NM_001164293.1 | NP_001157765.1 | 9 | 835 | |||
NM_001164294.1 | NP_001157766.1 | 9 | 814 | |||
NM_001164295.1 | NP_001157767.1 | 9 | 811 | |||
NM_024192.2 | NP_077154.2 | 10 | 967 | |||
GenBank mRNA | AB232359.1 | - | 8 | 766 | ||
AK003457.1 | - | 9 | 770 | |||
AK013884.1 | - | 9 | 826 | |||
BC003442.1 | - | 10 | 931 | 2650719 |