Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026044.3 | NP_080320.1 | 9 | 1439 | ||
GenBank mRNA | AK013297.1 | - | 9 | 1419 | ||
AK078448.1 | - | 9 | 1402 | |||
AK087998.1 | - | 8 | 1314 | |||
AK152000.1 | - | 8 | 1352 | |||
AK153537.1 | - | 8 | 1352 | |||
BC056962.1 | - | 7 | 2089 | |||
BC086486.1 | - | 9 | 1441 |