Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290636.1 | NP_001277565.1 | 24 | 3024 | ||
NM_026082.5 | NP_080358.3 | 23 | 2934 | |||
GenBank mRNA | BC086672.1 | - | 4 | 442 | 30647593 | |
BC094420.1 | - | 11 | 1220 | |||
CB246838.1 | - | 3 | 190 | |||
CD802819.1 | - | 4 | 586 |