Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adck3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163290.1 | NP_001156762.1 | 14 | 1689 | ||
NM_023341.3 | NP_075830.2 | 14 | 1787 | |||
GenBank mRNA | AJ278735.1 | - | 14 | 1667 | ||
AK004177.1 | - | 7 | 672 | |||
AK004791.1 | - | 14 | 1729 | |||
AK014605.2 | - | 14 | 1729 | |||
AK179063.1 | - | 56 | ||||
BC030937.1 | - | 14 | 1735 | 4922892 |